Assay Details
Target Gene Details
Entrez Gene ID: | 9946 |
Gene Name: | crystallin zeta like 1 |
Gene Aliases: |
4P11, QOH-1 |
Location: |
Chr.21:33589341-33641854 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CRYZL1 | NM_145858.2 | 11 | 1020 | NP_665857.2 |
AF029689.1 | 11 | 887 | AAD22381.1 | |
AF086550.1 | ||||
AF098798.1 | 11 | 887 | AAD20219.1 | |
AK001293.1 | 11 | 913 | BAA91605.1 | |
AK057604.1 | 11 | 1541 | ||
AK058071.1 | 6 | 569 | ||
AK315707.1 | 11 | 880 | ||
BC033023.2 | 11 | 874 | AAH33023.1 | |
BX647276.1 | 10 | 899 | ||
BX648547.1 | 12 | 1034 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |