Assay Details
Target Gene Details
Entrez Gene ID: | 128338 |
Gene Name: | DNA damage regulated autophagy modulator 2 |
Gene Aliases: |
CORD21, PRO180, TMEM77, WWFQ154 |
Location: |
Chr.1:111117332-111140216 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DRAM2 | NM_178454.4 | 9 | 1153 | NP_848549.3 |
XM_005270469.2 | 10 | 1211 | XP_005270526.1 | |
XM_005270470.1 | 10 | 1334 | XP_005270527.1 | |
XM_006710361.1 | 10 | 1171 | XP_006710424.1 | |
XM_006710362.1 | 10 | 1144 | XP_006710425.1 | |
XM_011540707.1 | 10 | 1152 | XP_011539009.1 | |
XM_011540708.1 | 10 | 1232 | XP_011539010.1 | |
XM_017000320.1 | 9 | 1095 | XP_016855809.1 | |
XM_017000321.1 | 9 | 954 | XP_016855810.1 | |
AK075350.1 | 5 | 599 | BAC11562.1 | |
AK309557.1 | ||||
AL833098.1 | 9 | 1521 | ||
AY336747.1 | 9 | 1031 | AAR02410.1 | |
AY358492.1 | 9 | 1016 | AAQ88856.1 | |
BC047025.1 | 8 | 968 | AAH47025.3 | |
BC091509.1 | 10 | 1245 | AAH91509.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |