Assay Details
Target Gene Details
Entrez Gene ID: | 817 |
Gene Name: | calcium/calmodulin dependent protein kinase II delta |
Gene Aliases: |
CAMKD |
Location: |
Chr.4:113451032-113761927 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830038 | Chr.4:113289857 - 113470695 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)