Assay Details
Target Gene Details
Entrez Gene ID: | 23542 |
Gene Name: | mitogen-activated protein kinase 8 interacting protein 2 |
Gene Aliases: |
IB-2, IB2, JIP2, PRKM8IPL |
Location: |
Chr.22:50600685-50613978 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAPK8IP2 | NM_012324.4 | NP_036456.1 | ||
XM_011530679.2 | XP_011528981.1 | |||
XM_011530680.2 | XP_011528982.1 | |||
XM_011530681.2 | XP_011528983.1 | |||
AF136382.1 | AAF00980.1 | |||
AF218778.1 | AAF32323.1 | |||
AK293580.1 | ||||
AK294265.1 | ||||
BC009940.2 | AAH09940.2 | |||
BC047527.1 | AAH47527.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv589224 | Chr.22:50589773 - 50639823 on Build GRCh38 | Loss | MAPK8IP2 CHKB-AS1 ARSA |
nsv1067172 | Chr.22:50586195 - 50677098 on Build GRCh38 | Gain | SHANK3 MAPK8IP2 CHKB-AS1 ARSA |
nsv1059234 | Chr.22:50560544 - 50617472 on Build GRCh38 | Gain | CPT1B CHKB SYCE3 MAPK8IP2 CHKB-CPT1B CHKB-AS1 |
nsv829340 | Chr.22:50505720 - 50603696 on Build GRCh38 | Gain | CPT1B CHKB SYCE3 MAPK8IP2 CHKB-CPT1B SCO2 KLHDC7B NCAPH2 TYMP LMF2 ODF3B CHKB-AS1 |
nsv829343 | Chr.22:50599349 - 50606643 on Build GRCh38 | Gain | MAPK8IP2 |
nsv834239 | Chr.22:50485317 - 50630215 on Build GRCh38 | Loss | CHKB-CPT1B MIOX NCAPH2 ODF3B CHKB-AS1 ARSA CPT1B CHKB SYCE3 MAPK8IP2 SCO2 KLHDC7B ADM2 TYMP LMF2 |
nsv955203 | Chr.22:50599072 - 50631372 on Build GRCh38 | Deletion | MAPK8IP2 ARSA |
nsv589223 | Chr.22:50539595 - 50619495 on Build GRCh38 | Loss | CPT1B CHKB SYCE3 MAPK8IP2 CHKB-CPT1B KLHDC7B CHKB-AS1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |