Assay Details
Target Gene Details
Entrez Gene ID: | 10054 |
Gene Name: | ubiquitin like modifier activating enzyme 2 |
Gene Aliases: |
ARX, HRIHFB2115, SAE2 |
Location: |
Chr.19:34428176-34469893 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UBA2 | NM_005499.2 | NP_005490.1 | ||
XM_005258404.3 | XP_005258461.2 | |||
XM_006722962.2 | XP_006723025.1 | |||
XM_011526304.2 | XP_011524606.1 | |||
XM_017026134.1 | XP_016881623.1 | |||
AF079566.1 | AAD23914.1 | |||
AF090384.1 | AAD12784.1 | |||
AF110957.1 | AAD24434.1 | |||
AK023120.1 | ||||
AK124730.1 | ||||
AK299514.1 | ||||
AK315521.1 | ||||
AL136905.1 | CAB66839.1 | |||
BC003153.1 | AAH03153.1 | |||
BG503066.1 | ||||
BT009781.1 | AAP88783.1 | |||
CR456756.1 | CAG33037.1 | |||
U35832.1 | AAC99992.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1059395 | Chr.19:34430094 - 34499592 on Build GRCh38 | Gain | UBA2 WTIP |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |