Assay Details
Target Gene Details
Entrez Gene ID: | 79143 |
Gene Name: | membrane bound O-acyltransferase domain containing 7 |
Gene Aliases: |
BB1, LENG4, LPIAT, LRC4, MBOA7, OACT7, hMBOA-7 |
Location: |
Chr.19:54173412-54190050 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MBOAT7 | NM_001146056.2 | NP_001139528.1 | ||
NM_001146082.2 | NP_001139554.1 | |||
NM_001146083.2 | NP_001139555.1 | |||
NM_024298.4 | NP_077274.3 | |||
XM_011527299.2 | XP_011525601.1 | |||
XM_011527300.2 | XP_011525602.1 | |||
XM_017027296.1 | XP_016882785.1 | |||
AK055908.1 | BAB71043.1 | |||
AK122789.1 | ||||
AK123290.1 | 1 | 1828 | ||
AK293199.1 | ||||
AK298689.1 | ||||
AK311493.1 | ||||
BC002512.2 | AAH02512.2 | |||
BC003164.1 | AAH03164.2 | |||
BC006309.2 | AAH06309.1 | |||
BC015857.1 | ||||
BU633996.1 | ||||
CB240899.1 | ||||
EU016381.1 | ||||
S82470.1 | AAB37433.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9751 | Chr.19:54158885 - 54193528 on Build GRCh38 | Gain | LENG1 TMC4 MBOAT7 TSEN34 |
nsv515623 | Chr.19:54102901 - 54179150 on Build GRCh38 | Loss | LENG1 TMC4 TFPT NDUFA3 CNOT3 MBOAT7 PRPF31 |
nsv953613 | Chr.19:54097214 - 54198733 on Build GRCh38 | Deletion | LENG1 TMC4 TFPT NDUFA3 CNOT3 MBOAT7 OSCAR TSEN34 PRPF31 |
esv2718812 | Chr.19:53184636 - 54322450 on Build GRCh38 | Deletion | CACNG8 MIR519A2 ZNF765 TMC4 ZNF845 MIR518E MIR519B ZNF677 LOC105372457 MIR521-2 MIR519C NLRP12 MIR372 MIR527 MIR520E MIR518C MIR519D MIR518D MIR935 PRKCG VN1R4 MYADM ZNF813 LILRB3 VSTM1 MIR512-2 MIR516B2 MIR520F MIR1283-1 DPRX CNOT3 MIR520A MIR371B CACNG6 LILRA5 RPS9 ZNF665 MIR516B1 MIR525 TARM1 MIR4752 MIR516A1 MIR518F LILRB5 MIR515-2 MIR1283-2 MIR519E BIRC8 MIR526B MIR520B MIR498 TPM3P9 ZNF818P MIR518A1 MIR520G LILRB2 MIR519A1 MIR1323 LILRA6 PRPF31 MIR512-1 VN1R2 LOC284379 MIR520C MIR517C MIR524 TFPT MIR526A2 MIR515-1 ZNF331 MIR517B ZNF761 FAM90A27P MIR521-1 MIR520D MIR518A2 MIR517A MIR526A1 ZNF525 NDUFA3 MIR516A2 MIR371A OSCAR MIR518B MIR373 MIR522 LENG1 CACNG7 MBOAT7 MIR520H TSEN34 MIR523 |
nsv510774 | Chr.19:54158011 - 54269511 on Build GRCh38 | Deletion | LENG1 TMC4 MBOAT7 LILRB5 TSEN34 LILRB3 RPS9 LILRA6 |
esv2758771 | Chr.19:54163847 - 54954956 on Build GRCh38 | Gain+Loss | LAIR2 TMC4 MIR8061 NCR1 CDC42EP5 LILRA4 FCAR LILRB2 LILRB4 LILRA6 LOC101928804 LOC102724300 LENG9 LILRB3 LILRA2 NLRP7 LENG8-AS1 KIR2DL1 LENG8 LILRB1 TTYH1 KIR3DX1 LILRA5 LILRP2 LILRA1 RPS9 KIR2DL4 KIR2DL3 KIR3DL3 MIR4752 LAIR1 MBOAT7 LILRB5 TSEN34 KIR2DS4 KIR3DL2 KIR3DL1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |