Assay Details
Target Gene Details
Entrez Gene ID: | 80333 |
Gene Name: | potassium voltage-gated channel interacting protein 4 |
Gene Aliases: |
CALP, KCHIP4 |
Location: |
Chr.4:20728520-21948801 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCNIP4 | NM_001035003.1 | 8 | 1245 | NP_001030175.1 |
NM_001035004.1 | 8 | 1166 | NP_001030176.1 | |
NM_025221.5 | 9 | 1182 | NP_079497.2 | |
NM_147181.3 | 8 | 1080 | NP_671710.1 | |
NM_147182.3 | 9 | 1192 | NP_671711.1 | |
NM_147183.3 | 8 | 999 | NP_671712.1 | |
XM_005248190.4 | 8 | 1095 | XP_005248247.1 | |
XM_011513882.2 | 8 | 3102 | XP_011512184.1 | |
XM_011513883.2 | 7 | 2971 | XP_011512185.1 | |
XM_011513884.2 | 7 | 2995 | XP_011512186.1 | |
XM_011513885.2 | 7 | 8291 | XP_011512187.1 | |
XM_011513886.2 | 7 | 3006 | XP_011512188.1 | |
XM_011513887.2 | 7 | 3771 | XP_011512189.1 | |
XM_011513889.2 | 7 | 2961 | XP_011512191.1 | |
XM_017008653.1 | 8 | 1268 | XP_016864142.1 | |
AK289661.1 | 8 | 1074 | ||
AK289922.1 | 9 | 1152 | ||
AK290998.1 | 8 | 1082 | ||
BC032520.1 | 8 | 1120 | AAH32520.1 | |
BX648990.1 | 8 | 1246 | ||
DQ148490.1 | 8 | 1245 | AAZ77807.1 | |
DQ148491.1 | 8 | 1166 | AAZ77808.1 |
Target Gene Details
Entrez Gene ID: | 133015 |
Gene Name: | PARK2 coregulated like |
Gene Aliases: |
C4orf28 |
Location: |
Chr.4:20696282-20757766 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PACRGL | NR_133933.1 | |||
XM_011513786.2 | 11 | 3733 | XP_011512088.1 | |
XM_011513805.2 | 8 | 3274 | XP_011512107.1 | |
XM_017007747.1 | 10 | 3488 | XP_016863236.1 | |
XM_017007748.1 | 10 | 3439 | XP_016863237.1 | |
XM_017007749.1 | 9 | 3354 | XP_016863238.1 | |
XM_017007751.1 | 9 | 3556 | XP_016863240.1 | |
XM_017007752.1 | 8 | 3403 | XP_016863241.1 | |
XM_017007753.1 | 8 | 3273 | XP_016863242.1 | |
XM_017007754.1 | 6 | 3330 | XP_016863243.1 | |
XM_017007755.1 | 6 | 3147 | XP_016863244.1 | |
XM_017007756.1 | 6 | 3140 | XP_016863245.1 | |
XM_017007757.1 | 6 | 3076 | XP_016863246.1 | |
XM_017007758.1 | 5 | 3025 | XP_016863247.1 | |
XM_017007759.1 | 5 | 2973 | XP_016863248.1 | |
AK098692.1 | BAC05381.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829876 | Chr.4:20637113 - 20833998 on Build GRCh38 | Gain | KCNIP4 PACRGL |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |