Assay Details
Target Gene Details
Entrez Gene ID: | 27071 |
Gene Name: | dual adaptor of phosphotyrosine and 3-phosphoinositides 1 |
Gene Aliases: |
BAM32 |
Location: |
Chr.4:99816824-99872290 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DAPP1 | NM_001306151.1 | NP_001293080.1 | ||
NM_014395.2 | NP_055210.2 | |||
XM_011531840.2 | XP_011530142.1 | |||
XM_011531842.1 | XP_011530144.1 | |||
XM_017008023.1 | XP_016863512.1 | |||
XM_017008024.1 | XP_016863513.1 | |||
AF161551.1 | AAF29038.1 | |||
AF163254.1 | AAD49697.1 | |||
AF178987.1 | AAF44351.1 | |||
AF186022.1 | AAF14578.1 | |||
AK301355.1 | ||||
BC012924.1 | AAH12924.1 | |||
DN995752.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |