Assay Details
Target Gene Details
Entrez Gene ID: | 4644 |
Gene Name: | myosin VA |
Gene Aliases: |
GS1, MYH12, MYO5, MYR12 |
Location: |
Chr.15:52307283-52529050 on Build GRCh38 |
Assay Gene Location: | Within Exon 43 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MYO5A | NM_000259.3 | 41 | 7288 | NP_000250.3 |
NM_001142495.1 | 40 | 7207 | NP_001135967.1 | |
XM_005254397.3 | 42 | 7195 | XP_005254454.1 | |
XM_011521606.2 | 42 | 7198 | XP_011519908.2 | |
XM_011521607.2 | 41 | 7189 | XP_011519909.2 | |
XM_011521608.2 | 41 | 7122 | XP_011519910.2 | |
XM_011521609.2 | 41 | 7116 | XP_011519911.2 | |
XM_011521610.2 | 40 | 7114 | XP_011519912.2 | |
XM_011521611.2 | 40 | 7041 | XP_011519913.2 | |
XM_011521612.2 | 39 | 7032 | XP_011519914.2 | |
XM_017022227.1 | 41 | 7090 | XP_016877716.1 | |
AB209505.1 | 33 | 5820 | BAD92742.1 | |
AB290180.1 | 40 | 6966 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv819139 | Chr.15:52311467 - 52313569 on Build GRCh38 | Loss | MYO5A |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |