Assay Details
Target Gene Details
Entrez Gene ID: | 59 |
Gene Name: | actin, alpha 2, smooth muscle, aorta |
Gene Aliases: |
AAT6, ACTSA, MYMY5 |
Location: |
Chr.10:88935074-88991397 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACTA2 | NM_001141945.2 | NP_001135417.1 | ||
NM_001320855.1 | NP_001307784.1 | |||
NM_001613.2 | NP_001604.1 | |||
AK300664.1 | ||||
AK313294.1 | ||||
AY692464.1 | ||||
BC017554.2 | AAH17554.1 | |||
BC093052.1 | AAH93052.1 | |||
CR536518.1 | CAG38756.1 | |||
J05192.1 | AAA51577.1 | |||
X13839.1 | CAA32064.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831943 | Chr.10:88905079 - 89106077 on Build GRCh38 | Gain | FAS-AS1 FAS MIR4679-2 STAMBPL1 ACTA2 ACTA2-AS1 MIR4679-1 |
nsv831942 | Chr.10:88804940 - 88970595 on Build GRCh38 | Gain | FAS STAMBPL1 ANKRD22 LIPM ACTA2 ACTA2-AS1 |
esv3624144 | Chr.10:88876937 - 89011636 on Build GRCh38 | Gain | FAS-AS1 FAS STAMBPL1 ACTA2 ACTA2-AS1 |
esv3624139 | Chr.10:88724980 - 89003307 on Build GRCh38 | Gain | FAS-AS1 FAS STAMBPL1 LIPK LIPN ANKRD22 LIPM ACTA2 ACTA2-AS1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |