Assay Details
Target Gene Details
Entrez Gene ID: | 2110 |
Gene Name: | electron transfer flavoprotein dehydrogenase |
Gene Aliases: |
ETFQO, MADD |
Location: |
Chr.4:158672101-158708713 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ETFDH | NM_001281737.1 | NP_001268666.1 | ||
NM_001281738.1 | NP_001268667.1 | |||
NM_004453.3 | NP_004444.2 | |||
AK293737.1 | ||||
AK304838.1 | ||||
AK314803.1 | ||||
BC011890.1 | AAH11890.1 | |||
BX538129.1 | CAD98030.1 | |||
BX957055.1 | ||||
DB221748.1 | ||||
EU093082.1 | ||||
EU093083.1 | ||||
S69232.1 | AAC60628.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |