Assay Details
Target Gene Details
Entrez Gene ID: | 9650 |
Gene Name: | mitochondrial fission regulator 1 |
Gene Aliases: |
CHPPR, FAM54A2 |
Location: |
Chr.8:65643873-65771252 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MTFR1 | NM_001145838.1 | NP_001139310.1 | ||
NM_001145839.1 | NP_001139311.1 | |||
NM_014637.3 | NP_055452.3 | |||
XM_006716484.2 | XP_006716547.2 | |||
XM_011517626.1 | XP_011515928.1 | |||
XM_011517627.2 | XP_011515929.1 | |||
AA015794.1 | ||||
AK298945.1 | ||||
AK304716.1 | ||||
AK316202.1 | ||||
BC036116.1 | AAH36116.1 | |||
BC043498.2 | AAH43498.1 | |||
BX647507.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |