Assay Details
Target Gene Details
Entrez Gene ID: | 196264 |
Gene Name: | myelin protein zero like 3 |
Gene Aliases: |
- |
Location: |
Chr.11:118226690-118252368 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MPZL3 | NM_001286152.1 | NP_001273081.1 | ||
NM_198275.2 | NP_938016.1 | |||
NR_104404.1 | ||||
NR_104405.1 | ||||
XM_006718775.2 | XP_006718838.1 | |||
XM_017017304.1 | XP_016872793.1 | |||
AK095399.1 | ||||
AK289390.1 | ||||
AK296951.1 | ||||
AK304292.1 | ||||
AK310966.1 | ||||
AY358626.1 | AAQ88989.1 | |||
BC113586.1 | ||||
BC143897.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv511 | Chr.11:118204437 - 118239426 on Build GRCh38 | Insertion | JAML MPZL3 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |