Assay Details
Target Gene Details
Entrez Gene ID: | 8639 |
Gene Name: | amine oxidase, copper containing 3 |
Gene Aliases: |
HPAO, SSAO, VAP-1, VAP1 |
Location: |
Chr.17:42851123-42858130 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AOC3 | NM_001277731.1 | NP_001264660.1 | ||
NM_001277732.1 | NP_001264661.1 | |||
NM_003734.3 | NP_003725.1 | |||
NR_102422.1 | ||||
XM_011525419.1 | XP_011523721.1 | |||
XM_011525420.2 | XP_011523722.1 | |||
AF067406.1 | AAC25170.1 | |||
AK025727.1 | ||||
AK315129.1 | ||||
BC050549.1 | AAH50549.1 | |||
DA592022.1 | ||||
DB250074.1 | ||||
JX020506.1 | ||||
U39447.1 | AAC50919.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv483009 | Chr.17:42829470 - 43006887 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)