Assay Details
Target Gene Details
Entrez Gene ID: | 387129 |
Gene Name: | neuropeptide S receptor 1 |
Gene Aliases: |
ASRT2, GPR154, GPRA, NPSR, PGR14, VRR1 |
Location: |
Chr.7:34658239-34878332 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NPSR1 | NM_001300933.1 | NP_001287862.1 | ||
NM_001300934.1 | NP_001287863.1 | |||
NM_001300935.1 | NP_001287864.1 | |||
NM_207172.1 | NP_997055.1 | |||
NM_207173.1 | NP_997056.1 | |||
AY310326.1 | AAQ76966.1 | |||
AY310327.1 | AAQ76967.1 | |||
AY310328.1 | AAQ76968.1 | |||
AY310330.1 | AAQ76970.1 | |||
AY310331.1 | AAQ76971.1 | |||
AY310332.1 | AAQ76972.1 | |||
BC132693.1 | ||||
BC132695.1 | ||||
BC144380.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv1080e201 | Chr.7:32621750 - 35008557 on Build GRCh38 | Deletion | DPY19L1P2 NT5C3A LINC00997 NPSR1-AS1 FLJ20712 BBS9 KBTBD2 DPY19L1 RP9P RP9 MIR550A2 FKBP9 MIR550B2 LOC100421336 MIR548N BMPER DPY19L1P1 ZNRF2P1 NPSR1 |
nsv1020423 | Chr.7:34807572 - 34850088 on Build GRCh38 | Gain | NPSR1-AS1 NPSR1 |
dgv418n21 | Chr.7:34849610 - 34884725 on Build GRCh38 | Loss | NPSR1 |
nsv1135982 | Chr.7:32794601 - 34893593 on Build GRCh38 | Deletion | FLJ20712 BBS9 KBTBD2 RP9P RP9 FKBP9 DPY19L1P2 LOC100421336 BMPER NT5C3A NPSR1-AS1 NPSR1 |
nsv436907 | Chr.7:32668474 - 34999437 on Build GRCh38 | Insertion | DPY19L1P2 NT5C3A LINC00997 NPSR1-AS1 FLJ20712 BBS9 KBTBD2 DPY19L1 RP9P RP9 MIR550A2 FKBP9 MIR550B2 LOC100421336 MIR548N BMPER DPY19L1P1 ZNRF2P1 NPSR1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |