Assay Details
Target Gene Details
Entrez Gene ID: | 2887 |
Gene Name: | growth factor receptor bound protein 10 |
Gene Aliases: |
GRB-IR, Grb-10, IRBP, MEG1, RSS |
Location: |
Chr.7:50590063-50793462 on Build GRCh38 |
Assay Gene Location: | Within Exon 32 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1016665 | Chr.7:50384854 - 51336827 on Build GRCh38 | Gain | DDC FIGNL1 GRB10 IKZF1 COBL DDC-AS1 |
nsv1015758 | Chr.7:50497706 - 50700456 on Build GRCh38 | Gain | DDC GRB10 DDC-AS1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |