Assay Details
Target Gene Details
Entrez Gene ID: | 121643 |
Gene Name: | forkhead box N4 |
Gene Aliases: |
- |
Location: |
Chr.12:109277978-109309406 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FOXN4 | NM_213596.2 | NP_998761.2 | ||
XM_011537922.2 | XP_011536224.1 | |||
XM_011537923.2 | XP_011536225.1 | |||
XM_017018818.1 | XP_016874307.1 | |||
XM_017018819.1 | XP_016874308.1 | |||
AF425596.1 | AAL23949.1 | |||
AF425597.1 | AAL23950.1 | |||
AK131519.1 | BAD18661.1 | |||
BC113073.1 | ||||
BC146825.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |