Assay Details
Target Gene Details
Entrez Gene ID: | 7048 |
Gene Name: | transforming growth factor beta receptor 2 |
Gene Aliases: |
AAT3, FAA3, LDS1B, LDS2, LDS2B, MFS2, RIIC, TAAD2, TGFR-2, TGFbeta-RII |
Location: |
Chr.3:30606490-30694142 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TGFBR2 | NM_001024847.2 | NP_001020018.1 | ||
NM_003242.5 | NP_003233.4 | |||
XM_011534045.2 | XP_011532347.1 | |||
AK300383.1 | ||||
AK314102.1 | ||||
AL554950.3 | ||||
CB988859.1 | ||||
D28131.1 | BAA05673.1 | |||
DB493906.1 | ||||
KC792576.1 | ||||
KC792577.1 | ||||
KC792578.1 | ||||
KC792579.1 | ||||
KC792580.1 | ||||
KU178359.1 | ||||
KU178360.1 | ||||
M85079.1 | AAA61164.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |