Assay Details
Target Gene Details
Entrez Gene ID: | 9049 |
Gene Name: | aryl hydrocarbon receptor interacting protein |
Gene Aliases: |
ARA9, FKBP16, FKBP37, SMTPHN, XAP-2, XAP2 |
Location: |
Chr.11:67483029-67491108 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AIP | NM_001302960.1 | 1 | 60 | NP_001289889.1 |
NM_003977.3 | 1 | 60 | NP_003968.3 | |
BG821308.1 | 1 | 55 | ||
HY065637.1 | 1 | 60 | ||
U78521.1 | AAB59004.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv468604 | Chr.11:67418529 - 67516420 on Build GRCh38 | Loss | MIR6752 TMEM134 RPS6KB2 PTPRCAP AIP CABP4 PITPNM1 CARNS1 CDK2AP2 CORO1B GPR152 |
esv2422195 | Chr.11:67466851 - 67859430 on Build GRCh38 | Duplication | CABP2 MIR6752 TMEM134 ACY3 AIP CDK2AP2 TBX10 ALDH3B2 C11orf72 NUDT8 DOC2GP PITPNM1 NDUFV1 GSTP1 FAM86C2P |
dgv1977n54 | Chr.11:67397024 - 67521123 on Build GRCh38 | Loss | CABP2 MIR6752 TMEM134 TBC1D10C AIP CABP4 CARNS1 CDK2AP2 CORO1B RPS6KB2 PTPRCAP RAD9A PITPNM1 PPP1CA GPR152 |
nsv368 | Chr.11:67479842 - 67509866 on Build GRCh38 | Insertion | MIR6752 AIP PITPNM1 CDK2AP2 |
dgv52n111 | Chr.11:67362651 - 67521123 on Build GRCh38 | Deletion | CABP2 MIR6752 TMEM134 TBC1D10C CLCF1 AIP CABP4 CARNS1 CDK2AP2 CORO1B RPS6KB2 PTPRCAP RAD9A LOC100130987 PITPNM1 PPP1CA GPR152 |
nsv521723 | Chr.11:67408221 - 67543787 on Build GRCh38 | Loss | CABP2 MIR6752 TMEM134 TBC1D10C AIP CABP4 CARNS1 CDK2AP2 CORO1B RPS6KB2 PTPRCAP PITPNM1 GPR152 |
nsv527056 | Chr.11:67439955 - 67526837 on Build GRCh38 | Loss | CABP2 MIR6752 TMEM134 AIP CABP4 PITPNM1 CDK2AP2 CORO1B GPR152 |
nsv825960 | Chr.11:67471173 - 67483362 on Build GRCh38 | Gain | AIP |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |