Assay Details
Target Gene Details
Entrez Gene ID: | 57689 |
Gene Name: | leucine rich repeat containing 4C |
Gene Aliases: |
NGL-1, NGL1 |
Location: |
Chr.11:40107066-41460212 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRRC4C | NM_001258419.1 | 7 | 1836 | NP_001245348.1 |
NM_020929.2 | 5 | 1667 | NP_065980.1 | |
NR_047673.1 | 3 | 2168 | ||
NR_047674.1 | 2 | 2115 | ||
XM_011520238.2 | 8 | 2624 | XP_011518540.1 | |
XM_011520239.2 | 9 | 2690 | XP_011518541.1 | |
XM_011520240.2 | 7 | 2544 | XP_011518542.1 | |
XM_011520241.2 | 4 | 3259 | XP_011518543.1 | |
XM_011520242.2 | 4 | 3245 | XP_011518544.1 | |
XM_011520243.2 | 7 | 2571 | XP_011518545.1 | |
XM_011520244.2 | 6 | 2491 | XP_011518546.1 | |
XM_017018070.1 | 8 | 2620 | XP_016873559.1 | |
XM_017018071.1 | 8 | 2610 | XP_016873560.1 | |
XM_017018072.1 | 8 | 2473 | XP_016873561.1 | |
XM_017018073.1 | 6 | 2332 | XP_016873562.1 | |
XM_017018074.1 | 4 | 3240 | XP_016873563.1 | |
XM_017018075.1 | 5 | 2279 | XP_016873564.1 | |
XM_017018076.1 | 3 | 3206 | XP_016873565.1 | |
XM_017018077.1 | 6 | 2359 | XP_016873566.1 | |
XM_017018078.1 | 5 | 3325 | XP_016873567.1 | |
XM_017018079.1 | 6 | 2487 | XP_016873568.1 | |
AB046800.1 | 2 | 3100 | BAB13406.1 | |
AK289646.1 | 3 | 1303 | ||
AY358293.1 | 2 | 1946 | AAQ88660.1 | |
BC041374.2 | 3 | 2168 | AAH41374.3 | |
DQ084201.1 | 7 | 1785 | AAY86708.1 | |
DQ084202.1 | 5 | 1804 | AAY86709.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv65n21 | Chr.11:40049473 - 40249892 on Build GRCh38 | Gain | LRRC4C |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |