Assay Details
Target Gene Details
Entrez Gene ID: | 55970 |
Gene Name: | G protein subunit gamma 12 |
Gene Aliases: |
- |
Location: |
Chr.1:67701466-67833779 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GNG12 | NM_018841.5 | 4 | 504 | NP_061329.3 |
XM_017001809.1 | 4 | 671 | XP_016857298.1 | |
XM_017001810.1 | 5 | 912 | XP_016857299.1 | |
XM_017001811.1 | 5 | 841 | XP_016857300.1 | |
AF119663.1 | 4 | 488 | AAF17220.1 | |
AF123766.1 | 4 | 495 | AAP97245.1 | |
AF365871.1 | 4 | 431 | AAK53385.1 | |
AK293101.1 | 5 | 445 | ||
AL832431.1 | 4 | 457 | CAH10645.1 | |
BC005940.1 | 4 | 485 | AAH05940.1 | |
BP236225.1 | 4 | 491 | ||
BP236699.1 | 4 | 504 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |