Assay Details
Target Gene Details
Entrez Gene ID: | 54207 |
Gene Name: | potassium two pore domain channel subfamily K member 10 |
Gene Aliases: |
K2p10.1, PPP1R97, TREK-2, TREK2 |
Location: |
Chr.14:88180108-88326912 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCNK10 | NM_021161.4 | 7 | 2397 | NP_066984.1 |
NM_138317.2 | 7 | 2406 | NP_612190.1 | |
NM_138318.2 | 7 | 2077 | NP_612191.1 | |
XM_011536840.2 | 6 | 1782 | XP_011535142.1 | |
AF279890.1 | 7 | 2412 | AAG15191.1 | |
AF385399.1 | 7 | 2078 | AAL95705.1 | |
AF385400.1 | 7 | 2160 | AAL95706.1 | |
AK290518.1 | ||||
EU978938.1 | ||||
EU978939.1 | ||||
EU978940.1 | ||||
EU978941.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |