Assay Details
Target Gene Details
Entrez Gene ID: | 4781 |
Gene Name: | nuclear factor I B |
Gene Aliases: |
CTF, HMGIC/NFIB, NF-I/B, NF1-B, NFI-B, NFI-RED, NFIB2, NFIB3 |
Location: |
Chr.9:14081843-14398983 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFIB | NM_001190737.1 | 11 | 6506 | NP_001177666.1 |
NM_001190738.1 | 9 | 6179 | NP_001177667.1 | |
NM_001282787.1 | 11 | 5642 | NP_001269716.1 | |
NM_005596.3 | 9 | 6284 | NP_005587.2 | |
XM_005251467.2 | 12 | 6715 | XP_005251524.1 | |
XM_005251468.4 | 12 | 6188 | XP_005251525.1 | |
XM_005251469.2 | 12 | 6688 | XP_005251526.1 | |
XM_005251470.2 | 10 | 6466 | XP_005251527.1 | |
XM_005251471.3 | 11 | 7550 | XP_005251528.1 | |
XM_006716773.2 | 12 | 6712 | XP_006716836.1 | |
XM_006716774.2 | 11 | 6626 | XP_006716837.1 | |
XM_006716775.2 | 10 | 6543 | XP_006716838.1 | |
XM_017014739.1 | 9 | 6374 | XP_016870228.1 | |
XM_017014740.1 | 9 | 6195 | XP_016870229.1 | |
XM_017014741.1 | 9 | 6312 | XP_016870230.1 | |
XM_017014742.1 | 9 | 7827 | XP_016870231.1 | |
AK094066.1 | 1 | 433 | ||
BX537698.1 | 1 | 3081 | ||
BX537707.1 | 1 | 2653 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1033764 | Chr.9:13764919 - 14226169 on Build GRCh38 | Loss | LINC00583 NFIB |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |