Assay Details
Target Gene Details
Entrez Gene ID: | 1267 |
Gene Name: | 2',3'-cyclic nucleotide 3' phosphodiesterase |
Gene Aliases: |
CNP1 |
Location: |
Chr.17:41966741-41977736 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CNP | NM_033133.4 | 4 | 4094 | NP_149124.3 |
AK098048.1 | 1 | 912 | ||
AK124861.1 | 1 | 1444 |
Target Gene Details
Entrez Gene ID: | 7266 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C7 |
Gene Aliases: |
DJ11, DJC7, TPR2, TTC2 |
Location: |
Chr.17:41976421-42021371 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DNAJC7 | NM_001144766.2 | 14 | 1659 | NP_001138238.1 |
NM_003315.3 | 14 | 1891 | NP_003306.3 | |
NR_029431.1 | 13 | 1802 | ||
XM_011525167.2 | 15 | 1821 | XP_011523469.1 | |
XM_011525168.2 | 15 | 1781 | XP_011523470.1 | |
XM_011525169.2 | 14 | 1633 | XP_011523471.1 | |
XM_017024994.1 | 15 | 1815 | XP_016880483.1 | |
AB209834.1 | 14 | 1577 | BAD93071.1 | |
AK291047.1 | 14 | 1592 | ||
AK296602.1 | 14 | 1652 | ||
AK298860.1 | 14 | 1607 | ||
AK316081.1 | ||||
BC003601.1 | 13 | 1514 | AAH03601.1 | |
BC011837.2 | 14 | 1610 | AAH11837.2 | |
BC033772.1 | 14 | 1636 | AAH33772.1 | |
BQ445927.1 | 1 | 206 | ||
BX647209.1 | 14 | 1847 | ||
U46571.1 | 14 | 1606 | AAB36872.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953897 | Chr.17:41966883 - 41977382 on Build GRCh38 | Deletion | DNAJC7 CNP |
nsv509658 | Chr.17:41927491 - 42002888 on Build GRCh38 | Insertion | DNAJC7 CNP TTC25 ACLY |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |