Assay Details
Target Gene Details
Entrez Gene ID: | 83892 |
Gene Name: | potassium channel tetramerization domain containing 10 |
Gene Aliases: |
BTBD28, MSTP028, ULRO61, hBACURD3 |
Location: |
Chr.12:109448655-109477544 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCTD10 | NM_001317395.1 | NP_001304324.1 | ||
NM_001317399.1 | NP_001304328.1 | |||
NM_031954.4 | NP_114160.1 | |||
NR_133897.1 | ||||
NR_133898.1 | ||||
AB209358.1 | 1 | 3362 | BAD92595.1 | |
AF113208.1 | AAG39279.1 | |||
AK027543.1 | BAB55188.1 | |||
AK056912.1 | ||||
AK092170.1 | ||||
AK123733.1 | ||||
AK222548.1 | BAD96268.1 | |||
AK309783.1 | ||||
AY205299.1 | AAO47716.1 | |||
AY597809.1 | AAT09002.1 | |||
BC040062.1 | AAH40062.1 | |||
BX333975.2 | ||||
CX785497.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3580463 | Chr.12:109455759 - 109457363 on Build GRCh38 | Loss | KCTD10 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |