Assay Details
Target Gene Details
Entrez Gene ID: | 25897 |
Gene Name: | ring finger protein 19A, RBR E3 ubiquitin protein ligase |
Gene Aliases: |
RNF19 |
Location: |
Chr.8:100257059-100336218 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RNF19A | NM_001280539.1 | 11 | 3654 | NP_001267468.1 |
NM_015435.4 | 11 | 3685 | NP_056250.3 | |
NM_183419.3 | 10 | 3761 | NP_904355.1 | |
XM_005250853.3 | 10 | 3723 | XP_005250910.1 | |
XM_011516956.2 | 9 | 3584 | XP_011515258.1 | |
XM_017013302.1 | 11 | 3846 | XP_016868791.1 | |
XM_017013303.1 | 11 | 3756 | XP_016868792.1 | |
XM_017013304.1 | 9 | 3541 | XP_016868793.1 | |
XM_017013305.1 | 8 | 3241 | XP_016868794.1 | |
AB029316.1 | 11 | 3685 | BAB39353.1 | |
AK023455.1 | 1 | 1361 | BAB14581.1 | |
AK027070.1 | 5 | 2104 | BAB15647.1 | |
AL110253.1 | 1 | 1159 | CAB53700.1 | |
AL122096.1 | 3 | 1801 | CAB59264.1 | |
BC043648.1 | 1 | 1346 | AAH43648.1 | |
BQ003778.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv820040 | Chr.8:100257101 - 100259800 on Build GRCh38 | Loss | RNF19A |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |