Assay Details
Target Gene Details
Entrez Gene ID: | 64895 |
Gene Name: | poly(A) polymerase gamma |
Gene Aliases: |
- |
Location: |
Chr.2:60756230-60802086 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PAPOLG | NM_022894.3 | 1 | 140 | NP_075045.2 |
XM_005264500.3 | 1 | 140 | XP_005264557.1 | |
XM_005264501.1 | XP_005264558.1 | |||
XM_011533041.1 | XP_011531343.1 | |||
XM_017004723.1 | 1 | 114 | XP_016860212.1 | |
XM_017004724.1 | 1 | 114 | XP_016860213.1 | |
AF312211.1 | 1 | 122 | AAK83701.1 | |
AK021867.1 | 1 | 76 | BAB13919.1 | |
AK314663.1 | 1 | 104 | ||
BC016738.1 | 1 | 111 | ||
BC110906.1 | 1 | 79 | AAI10907.1 | |
BC111701.1 | AAI11702.1 | |||
BC113747.1 | AAI13748.1 | |||
BX647824.1 | ||||
DB082179.1 | 1 | 140 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv3859n100 | Chr.2:60754968 - 60868884 on Build GRCh38 | Gain | PAPOLG LINC01185 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |