Assay Details
Target Gene Details
Entrez Gene ID: | 283870 |
Gene Name: | BRICHOS domain containing 5 |
Gene Aliases: |
C16orf79 |
Location: |
Chr.16:2209253-2213037 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BRICD5 | NM_182563.3 | 1 | 199 | NP_872369.2 |
XM_005255263.4 | 1 | 2168 | XP_005255320.1 | |
AY597811.1 | AAT09004.1 | |||
BC039154.2 | 1 | 141 | AAH39154.1 | |
DN992777.1 | 1 | 207 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv571200 | Chr.16:2209190 - 2244889 on Build GRCh38 | Loss | ECI1 BRICD5 DNASE1L2 MLST8 PGP E4F1 |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | SNORD60 METTL26 TBL3 HN1L RHOT2 C1QTNF8 SNORA78 RPL23AP5 LOC105371045 TMEM8A RNPS1 FBXL16 ERVK13-1 FLJ42627 LINC00235 LOC105371049 SYNGR3 PRSS29P BAIAP3 MIR3176 PIGQ WASIR2 AMDHD2 LOC105371184 MLST8 CLCN7 LOC100134368 KCTD5 NPRL3 CCDC154 CCNF TSC2 DNASE1L2 ARHGDIG LOC105371038 LOC100287175 RGS11 HS3ST6 MRPS34 UBE2I MIR3677 SOX8 ATP6V0C CEMP1 TPSG1 RPS2 BRICD5 MAPK8IP3 LOC105371046 MIR6511B1 HBA2 RAB11FIP3 HAGH SSTR5 MIR3180-5 LOC105371047 SSTR5-AS1 ZNF598 HAGHL POLR3K LOC107984876 FAM234A PDPK1 PDIA2 MIR6859-4 RAB26 IFT140 TSR3 ECI1 SNHG9 NPW IGFALS GNG13 LMF1 MIR662 LOC729652 RHBDF1 C16orf91 TPSB2 CASKIN1 MIR6767 LOC101929280 DDX11L10 DECR2 MIR4516 LUC7L NTHL1 ABCA3 TRAF7 RAB40C NDUFB10 LOC106660606 CAPN15 EME2 SPSB3 HBA1 SNRNP25 CRAMP1 TPSAB1 RPUSD1 SNORA10 NME4 HBQ1 TPSD1 WFIKKN1 C16orf59 TELO2 RNF151 MIR940 MRPL28 SLC9A3R2 MIR5587 CCDC78 PGP MSRB1 PKD1 PRSS27 TMEM204 LOC652276 PTX4 GFER MIR4717 LOC107987233 MIR3178 NUBP2 WDR90 TBC1D24 METRN SNHG19 PRR25 NHLRC4 FAM173A MPG MSLN CHTF18 FAHD1 AXIN1 HBZ JMJD8 NOXO1 NARFL E4F1 NTN3 ABCA17P MCRIP2 RPL3L STUB1 MIR1225 PRR35 UNKL LINC00254 WDR24 MEIOB HBM LMF1-AS1 MIR3177 GNPTG RHBDL1 SNORA64 CACNA1H NME3 MIR6768 |
nsv471072 | Chr.16:1971892 - 2317508 on Build GRCh38 | Loss | SNORD60 MIR3677 TRAF7 TBL3 LOC106660606 BRICD5 MIR6511B1 RNPS1 MIR3180-5 NOXO1 ZNF598 LOC105371049 SYNGR3 E4F1 MIR940 RAB26 ECI1 NPW SLC9A3R2 MLST8 MIR1225 PGP PKD1 GFER MIR4717 TSC2 DNASE1L2 CASKIN1 SNHG19 MIR4516 NTHL1 ABCA3 |
dgv4861n54 | Chr.16:2083900 - 2246850 on Build GRCh38 | Loss | SNORD60 RAB26 TRAF7 ECI1 BRICD5 MIR6511B1 MLST8 MIR1225 PGP PKD1 MIR3180-5 TSC2 DNASE1L2 CASKIN1 LOC105371049 SNHG19 MIR4516 E4F1 |
dgv4863n54 | Chr.16:2141733 - 2274302 on Build GRCh38 | Loss | SNORD60 MIR3677 RAB26 TRAF7 ECI1 LOC106660606 BRICD5 MLST8 PGP RNPS1 DNASE1L2 CASKIN1 SNHG19 E4F1 MIR940 |
nsv1711 | Chr.16:2199093 - 2232650 on Build GRCh38 | Insertion | BRICD5 MLST8 PGP E4F1 |
nsv457351 | Chr.16:2156590 - 2274302 on Build GRCh38 | Loss | MIR3677 TRAF7 ECI1 LOC106660606 BRICD5 DNASE1L2 CASKIN1 MLST8 PGP RNPS1 E4F1 MIR940 |
nsv1052260 | Chr.16:2074546 - 2629655 on Build GRCh38 | Gain | SNORD60 MIR3677 ATP6V0C CEMP1 TRAF7 LOC106660606 BRICD5 MIR6511B1 RNPS1 MIR3180-5 LOC105371049 C16orf59 PDPK1 E4F1 NTN3 MIR940 ABCA17P AMDHD2 RAB26 ECI1 MLST8 MIR1225 LOC729652 PGP PKD1 LOC652276 MIR4717 CCNF MIR3178 TSC2 DNASE1L2 CASKIN1 TBC1D24 MIR6767 SNHG19 MIR4516 MIR6768 ABCA3 |
nsv952902 | Chr.16:2143400 - 2339999 on Build GRCh38 | Deletion | SNORD60 MIR3677 RAB26 TRAF7 ECI1 LOC106660606 BRICD5 MLST8 PGP RNPS1 MIR4717 DNASE1L2 CASKIN1 SNHG19 E4F1 MIR940 ABCA3 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |