Assay Details
Target Gene Details
Entrez Gene ID: | 81544 |
Gene Name: | glycerophosphodiester phosphodiesterase domain containing 5 |
Gene Aliases: |
GDE2, PP1665 |
Location: |
Chr.11:75434640-75525916 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GDPD5 | NM_030792.6 | 17 | 2676 | NP_110419.5 |
XM_006718697.3 | 18 | 3282 | XP_006718760.1 | |
XM_011545276.2 | 19 | 3071 | XP_011543578.1 | |
XM_011545277.2 | 19 | 3459 | XP_011543579.1 | |
XM_011545278.2 | 19 | 3522 | XP_011543580.1 | |
XM_011545279.2 | 17 | 3041 | XP_011543581.1 | |
XM_011545280.1 | 16 | 2374 | XP_011543582.1 | |
XM_011545281.2 | 12 | 2194 | XP_011543583.1 | |
XM_011545285.2 | 13 | 4130 | XP_011543587.1 | |
XM_011545286.1 | 12 | 1930 | XP_011543588.1 | |
XM_011545287.1 | 12 | 1927 | XP_011543589.1 | |
AF217989.1 | 5 | 1011 | AAG17232.1 | |
AF289614.1 | 1 | 117 | AAL55798.1 | |
AF318351.1 | 17 | 2532 | AAL55858.1 | |
AF318377.1 | 17 | 2945 | AAL55884.1 | |
AF451987.1 | 10 | 1690 | AAP97686.1 | |
AK074848.1 | 11 | 1764 | BAC11242.1 | |
AK091658.1 | 2 | 2433 | ||
AK098808.1 | 12 | 1927 | BAC05418.1 | |
AL833941.1 | 18 | 2713 | CAD38796.2 | |
AY358477.1 | 12 | 1943 | AAQ88841.1 | |
BC018771.1 | 18 | 2724 | AAH18771.1 | |
BC020203.1 | 1 | 531 | AAH20203.1 | |
BC029416.1 | 1 | 287 | AAH29416.1 | |
BC033391.1 | 13 | 3666 | AAH33391.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |