Assay Details
Target Gene Details
Entrez Gene ID: | 64852 |
Gene Name: | terminal uridylyl transferase 1, U6 snRNA-specific |
Gene Aliases: |
PAPD2, RBM21, STARPAP, TUTase, URLC6 |
Location: |
Chr.11:62575045-62591637 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TUT1 | NM_022830.2 | NP_073741.2 | ||
AB101208.1 | ||||
AK025920.1 | BAB15282.1 | |||
AK026000.1 | BAB15314.1 | |||
AK292610.1 | ||||
BC005013.1 | AAH05013.2 | |||
BC110910.1 | AAI10911.1 | |||
BC128263.1 | ||||
KU178756.1 | ||||
KU178757.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3584599 | Chr.11:62495270 - 62630578 on Build GRCh38 | Gain | GANAB ROM1 EEF1G MTA2 MIR6747 AHNAK EML3 TUT1 B3GAT3 |
nsv832180 | Chr.11:62434133 - 62597356 on Build GRCh38 | Loss | EEF1G MTA2 MIR6747 AHNAK TUT1 |
nsv825939 | Chr.11:62558611 - 62643105 on Build GRCh38 | Gain | GANAB ROM1 EEF1G MTA2 MIR6747 EML3 TUT1 B3GAT3 |
nsv832182 | Chr.11:62528103 - 62695376 on Build GRCh38 | Gain+Loss | LRRN4CL LBHD1 INTS5 MTA2 MIR6747 AHNAK BSCL2 TUT1 B3GAT3 METTL12 UBXN1 UQCC3 GANAB ROM1 EEF1G HNRNPUL2-BSCL2 EML3 SNORA57 C11orf98 |
nsv555174 | Chr.11:62566624 - 62729047 on Build GRCh38 | Gain | LRRN4CL LBHD1 INTS5 MTA2 GNG3 MIR6747 TTC9C BSCL2 TUT1 B3GAT3 HNRNPUL2 METTL12 UBXN1 UQCC3 GANAB ROM1 EEF1G HNRNPUL2-BSCL2 EML3 SNORA57 C11orf98 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |