Assay Details
Target Gene Details
Entrez Gene ID: | 5518 |
Gene Name: | protein phosphatase 2 scaffold subunit Aalpha |
Gene Aliases: |
MRD36, PP2A-Aalpha, PP2AAALPHA, PR65A |
Location: |
Chr.19:52189802-52226425 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPP2R1A | NM_014225.5 | NP_055040.2 | ||
NR_033500.1 | ||||
XM_017026929.1 | XP_016882418.1 | |||
AK090488.1 | ||||
AK091387.1 | BAC03652.1 | |||
AK130000.1 | ||||
AK290593.1 | ||||
AK291932.1 | ||||
AK293174.1 | ||||
AK293493.1 | ||||
AK299012.1 | ||||
AK307743.1 | ||||
AK315700.1 | ||||
AU128747.1 | ||||
BC001537.2 | AAH01537.1 | |||
CR450340.1 | CAG29336.1 | |||
DB246691.1 | ||||
DC398468.1 | ||||
J02902.1 | AAA36399.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3583508 | Chr.19:52201672 - 52217804 on Build GRCh38 | Loss | PPP2R1A |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |