Assay Details
Target Gene Details
Entrez Gene ID: | 171017 |
Gene Name: | zinc finger protein 384 |
Gene Aliases: |
CAGH1, CAGH1A, CIZ, ERDA2, NMP4, NP, TNRC1 |
Location: |
Chr.12:6666477-6689572 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832322 | Chr.12:6575884 - 6765914 on Build GRCh38 | Loss | PIANP ING4 CHD4 LPAR5 SCARNA11 ZNF384 COPS7A MLF2 ACRBP |
nsv832323 | Chr.12:6662233 - 6856559 on Build GRCh38 | Loss | PIANP LAG3 CDCA3 GPR162 USP5 MLF2 CD4 ING4 PTMS P3H3 GNB3 ZNF384 COPS7A |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |