Assay Details
Target Gene Details
Entrez Gene ID: | 387338 |
Gene Name: | NOP2/Sun RNA methyltransferase family member 4 |
Gene Aliases: |
SHTAP |
Location: |
Chr.1:46340177-46365152 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NSUN4 | NM_001256127.1 | 6 | 2446 | NP_001243056.1 |
NM_001256128.1 | 6 | 2078 | NP_001243057.1 | |
NM_199044.3 | 6 | 2501 | NP_950245.2 | |
NR_045789.1 | 7 | 2676 | ||
NR_045790.1 | 5 | 2346 | ||
NR_045791.1 | 5 | 2164 | ||
BX648592.1 | 5 | 1735 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829748 | Chr.1:46258836 - 46410450 on Build GRCh38 | Loss | NSUN4 FAAH UQCRH LRRC41 RAD54L |
esv2761743 | Chr.1:45874186 - 46561071 on Build GRCh38 | Gain | UQCRH FAAHP1 LURAP1 MAST2 MKNK1-AS1 LINC01398 KNCN NSUN4 POMGNT1 FAAH MKNK1 DMBX1 PIK3R3 LRRC41 LOC101929626 TSPAN1 RAD54L |
nsv546162 | Chr.1:46341925 - 46405089 on Build GRCh38 | Loss | NSUN4 FAAH |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |