Assay Details
Target Gene Details
Entrez Gene ID: | 10850 |
Gene Name: | C-C motif chemokine ligand 27 |
Gene Aliases: |
ALP, CTACK, CTAK, ESKINE, ILC, PESKY, SCYA27 |
Location: |
Chr.9:34661883-34662692 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCL27 | NM_006664.3 | NP_006655.1 | ||
AB010445.1 | BAA87046.1 | |||
AF082393.1 | AAD41238.1 | |||
AJ243542.1 | CAB46983.1 | |||
BC148263.1 | ||||
BM719357.1 | ||||
CA433929.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3620335 | Chr.9:33871811 - 34738198 on Build GRCh38 | Gain | SNORD121B C9orf24 NUDT2 ENHO DCAF12 UBAP2 CCL27 FAM219A SIGMAR1 UBAP1 UBE2R2 CCL21 RPP25L LOC730098 SNORD121A ARID3C KIAA1161 FAM205A CNTFR-AS1 CCL19 DCTN3 CNTFR LOC114224 IL11RA KIF24 DNAI1 GALT |
nsv1161857 | Chr.9:34537279 - 34669174 on Build GRCh38 | Deletion | LOC730098 DCTN3 CNTFR IL11RA SIGMAR1 ARID3C CNTFR-AS1 RPP25L CCL27 GALT |
dgv12659n54 | Chr.9:34552439 - 34663141 on Build GRCh38 | Loss | DCTN3 CNTFR IL11RA SIGMAR1 ARID3C CNTFR-AS1 RPP25L CCL27 GALT |
nsv831544 | Chr.9:34650003 - 34818951 on Build GRCh38 | Gain | PHF24 LOC730098 CCL19 IL11RA FAM205A CCL21 CCL27 GALT |
nsv818699 | Chr.9:34535211 - 34685943 on Build GRCh38 | Gain | LOC730098 DCTN3 CNTFR IL11RA SIGMAR1 ARID3C CNTFR-AS1 RPP25L CCL27 GALT |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |