Assay Details
Target Gene Details
Entrez Gene ID: | 105370757 |
Gene Name: | uncharacterized LOC105370757 |
Gene Aliases: |
- |
Location: |
Chr.15:32665702-32673065 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105370757 | NR_135505.1 | |||
BX088994.1 |
Target Gene Details
Entrez Gene ID: | 6447 |
Gene Name: | secretogranin V |
Gene Aliases: |
7B2, P7B2, SGNE1, SgV |
Location: |
Chr.15:32641669-32697097 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SCG5 | NM_001144757.1 | NP_001138229.1 | ||
NM_003020.3 | NP_003011.1 | |||
AK314965.1 | ||||
BC005349.1 | AAH05349.1 | |||
BC093053.1 | AAH93053.1 | |||
BP377248.1 | ||||
CR541824.1 | CAG46623.1 | |||
Y00757.1 | CAA68726.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv456785 | Chr.15:32633875 - 33060788 on Build GRCh38 | Gain | FMN1 GREM1 LOC100131315 SCG5 ARHGAP11A LOC105370757 |
nsv428302 | Chr.15:31646275 - 32694378 on Build GRCh38 | Gain+Loss | GOLGA8N LOC100653133 CHRNA7 LOC100996255 WHAMMP1 LOC105370940 LOC101060588 ULK4P1 SCG5 GOLGA8K LOC101928042 ARHGAP11A LOC105370757 OTUD7A GOLGA8O |
esv2760023 | Chr.15:31474097 - 32694378 on Build GRCh38 | Gain+Loss | GOLGA8N LOC100653133 CHRNA7 LOC100996255 WHAMMP1 LOC105370940 LOC101060588 ULK4P1 SCG5 GOLGA8K LOC101928042 ARHGAP11A LOC105370757 OTUD7A GOLGA8O |
nsv1052620 | Chr.15:32612402 - 34538731 on Build GRCh38 | Gain | PGBD4 EMC7 GREM1 SLC12A6 KATNBL1 NOP10 NUTM1 GOLGA8B TMCO5B EMC4 CHRM5 RYR3 FMN1 AVEN MIR1233-1 LOC100131315 SCG5 LOC101928134 GOLGA8A LOC101928042 ARHGAP11A LOC105370757 MIR1233-2 LPCAT4 |
nsv483039 | Chr.15:32574635 - 32751321 on Build GRCh38 | Gain | GOLGA8N GREM1 LOC100131315 SCG5 LOC101928042 ARHGAP11A LOC105370757 LOC100996255 |
esv3892661 | Chr.15:32393132 - 32776080 on Build GRCh38 | Gain | GOLGA8N GREM1 LOC100653133 LOC100996255 WHAMMP1 FMN1 LOC101060588 ULK4P1 LOC100131315 SCG5 GOLGA8K LOC101928042 ARHGAP11A LOC105370757 GOLGA8O |
esv33337 | Chr.15:31689863 - 34575097 on Build GRCh38 | Gain+Loss | EMC7 SLC12A6 LOC100653133 NUTM1 GOLGA8B TMCO5B CHRM5 FMN1 LOC105370940 ULK4P1 AVEN MIR1233-1 SCG5 LOC101928134 GOLGA8A GOLGA8K LOC105370757 GOLGA8O PGBD4 GOLGA8N GREM1 KATNBL1 NOP10 CHRNA7 EMC4 LOC100996255 WHAMMP1 RYR3 LOC101060588 LOC100131315 LOC101928042 ARHGAP11A MIR1233-2 LPCAT4 |
nsv469587 | Chr.15:32529739 - 32694378 on Build GRCh38 | Loss | GOLGA8N SCG5 LOC101928042 ARHGAP11A LOC105370757 LOC100996255 WHAMMP1 |
esv2422296 | Chr.15:32506737 - 32929014 on Build GRCh38 | Duplication | GOLGA8N FMN1 GREM1 LOC100131315 SCG5 LOC101928042 ARHGAP11A LOC105370757 LOC100996255 WHAMMP1 |
dgv4456n54 | Chr.15:32604149 - 34557061 on Build GRCh38 | Gain | EMC7 SLC12A6 NUTM1 GOLGA8B TMCO5B CHRM5 FMN1 AVEN MIR1233-1 SCG5 LOC101928134 GOLGA8A LOC105370757 PGBD4 GOLGA8N GREM1 KATNBL1 NOP10 EMC4 RYR3 LOC100131315 LOC101928042 ARHGAP11A MIR1233-2 LPCAT4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |