Assay Details
Target Gene Details
Entrez Gene ID: | 54876 |
Gene Name: | DDB1 and CUL4 associated factor 16 |
Gene Aliases: |
C4orf30 |
Location: |
Chr.4:17798236-17810761 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DCAF16 | NM_017741.3 | 3 | 1154 | NP_060211.3 |
XM_005248169.3 | 3 | 1142 | XP_005248226.1 | |
XM_005248171.3 | 2 | 1038 | XP_005248228.1 | |
XM_006713966.3 | 4 | 1313 | XP_006714029.1 | |
XM_017008327.1 | 2 | 2607 | XP_016863816.1 | |
XM_017008328.1 | 2 | 2592 | XP_016863817.1 | |
AK000287.1 | 1 | 276 | BAA91056.1 | |
AK056116.1 | 3 | 1154 | ||
BC050697.1 | 2 | 815 | ||
BC068025.1 | 1 | 453 | ||
BC101716.1 | 1 | 164 | AAI01717.1 | |
BC101718.1 | 1 | 164 | AAI01719.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |