Assay Details
Target Gene Details
Entrez Gene ID: | 23659 |
Gene Name: | phospholipase A2 group XV |
Gene Aliases: |
ACS, GXVPLA2, LLPL, LPLA2, LYPLA3 |
Location: |
Chr.16:68245344-68261062 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLA2G15 | NM_012320.3 | 6 | 856 | NP_036452.1 |
XM_005255886.3 | 6 | 704 | XP_005255943.1 | |
XM_011522979.2 | 7 | 925 | XP_011521281.1 | |
XM_011522980.2 | 7 | 796 | XP_011521282.2 | |
AB017494.1 | 6 | 805 | BAA76877.1 | |
AK001705.1 | 6 | 817 | ||
AK074828.1 | 6 | 856 | BAC11233.1 | |
AK127898.1 | 2 | 1256 | ||
AK222790.1 | 6 | 815 | BAD96510.1 | |
AK296263.1 | 6 | 702 | ||
AK298494.1 | 2 | 227 | ||
AK300596.1 | 5 | 614 | ||
AL110209.1 | 6 | 837 | CAB53675.1 | |
AL389957.1 | 2 | 113 | CAB97531.1 | |
AY358425.1 | 6 | 793 | AAQ88791.1 | |
BC011640.2 | 3 | 370 | AAH11640.2 | |
BC062605.1 | 6 | 820 | AAH62605.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |