Assay Details
Target Gene Details
Entrez Gene ID: | 3728 |
Gene Name: | junction plakoglobin |
Gene Aliases: |
ARVD12, CTNNG, DP3, DPIII, PDGB, PKGB |
Location: |
Chr.17:41754607-41786853 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
JUP | NM_002230.2 | 10 | 1817 | NP_002221.1 |
NM_021991.2 | 10 | 1817 | NP_068831.1 | |
XM_006721873.2 | 11 | 2067 | XP_006721936.1 | |
XM_006721874.2 | 10 | 1980 | XP_006721937.1 | |
XM_006721875.1 | 10 | 1899 | XP_006721938.1 | |
XM_006721878.1 | 10 | 1880 | XP_006721941.1 | |
XM_011524753.2 | 11 | 2103 | XP_011523055.1 | |
XM_011524755.1 | 10 | 1812 | XP_011523057.1 | |
XM_011524756.1 | 10 | 1710 | XP_011523058.1 | |
XM_011524757.2 | 10 | 1977 | XP_011523059.1 | |
XM_011524758.1 | 10 | 1706 | XP_011523060.1 | |
XM_017024588.1 | 10 | 1799 | XP_016880077.1 | |
XM_017024589.1 | 10 | 1800 | XP_016880078.1 | |
XM_017024590.1 | 10 | 1706 | XP_016880079.1 | |
AK226051.1 | 10 | 1794 | ||
AY243535.1 | 9 | 1690 | AAO85780.1 | |
BC000441.2 | 10 | 1803 | AAH00441.2 | |
BC011865.2 | 10 | 1961 | AAH11865.1 | |
KU178155.1 | 9 | 1684 | ||
KU178156.1 | 4 | 451 | ||
M23410.1 | 10 | 1800 | AAA64895.1 | |
Z68228.1 | 10 | 1803 | CAA92522.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |