Assay Details
Target Gene Details
Entrez Gene ID: | 79872 |
Gene Name: | Cbl proto-oncogene like 1 |
Gene Aliases: |
HAKAI, RNF188 |
Location: |
Chr.7:107743497-107764997 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CBLL1 | NM_001284291.1 | 6 | 1056 | NP_001271220.1 |
NM_024814.3 | 6 | 1059 | NP_079090.2 | |
NR_024199.2 | 7 | 1132 | ||
XM_011516580.2 | 7 | 1329 | XP_011514882.1 | |
XM_017012643.1 | 6 | 860 | XP_016868132.1 | |
XM_017012644.1 | 6 | 1250 | XP_016868133.1 | |
XM_017012645.1 | 7 | 1184 | XP_016868134.1 | |
AK026762.1 | 6 | 615 | BAB15544.1 | |
AK293352.1 | 6 | 888 | ||
AK307992.1 | 6 | 615 | ||
BC027460.2 | 5 | 584 | AAH27460.2 | |
BC130529.1 | 6 | 611 | ||
BC130531.1 | 6 | 611 | ||
BC144176.1 | 6 | 608 | ||
BX348716.2 | 2 | 197 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |