Assay Details
Target Gene Details
Entrez Gene ID: | 64699 |
Gene Name: | transmembrane protease, serine 3 |
Gene Aliases: |
DFNB10, DFNB8, ECHOS1, TADG12 |
Location: |
Chr.21:42371887-42396846 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMPRSS3 | NM_001256317.1 | NP_001243246.1 | ||
NM_024022.2 | NP_076927.1 | |||
NM_032404.2 | NP_115780.1 | |||
NR_046020.1 | ||||
AB038157.1 | BAB20077.1 | |||
AB038158.1 | BAB20078.1 | |||
AB038159.1 | BAB20079.1 | |||
AF201380.1 | AAG37012.1 | |||
AI251446.1 | ||||
AK131212.1 | ||||
AK172842.1 | BAD18806.1 | |||
AY358458.1 | 13 | 2314 | AAQ88823.1 | |
AY633572.1 | AAT66641.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |