Assay Details
Target Gene Details
Entrez Gene ID: | 84658 |
Gene Name: | adhesion G protein-coupled receptor E3 |
Gene Aliases: |
EMR3 |
Location: |
Chr.19:14600175-14674918 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADGRE3 | AL832663.1 | 16 | 2186 |
Target Gene Details
Entrez Gene ID: | 388512 |
Gene Name: | C-type lectin domain family 17 member A |
Gene Aliases: |
- |
Location: |
Chr.19:14580777-14612035 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 15 - Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CLEC17A | NM_001204118.1 | NP_001191047.1 | ||
NM_207390.3 | NP_997273.3 | |||
NR_109784.1 | ||||
NR_109785.1 | ||||
XM_017026785.1 | XP_016882274.1 | |||
XM_017026786.1 | XP_016882275.1 | |||
XM_017026787.1 | XP_016882276.1 | |||
XM_017026788.1 | XP_016882277.1 | |||
XM_017026789.1 | XP_016882278.1 | |||
XM_017026790.1 | XP_016882279.1 | |||
XM_017026791.1 | XP_016882280.1 | |||
XM_017026792.1 | XP_016882281.1 | |||
XM_017026793.1 | XP_016882282.1 | |||
BC140848.1 | ||||
BC144665.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |