Assay Details
Target Gene Details
Entrez Gene ID: | 10873 |
Gene Name: | malic enzyme 3 |
Gene Aliases: |
NADP-ME |
Location: |
Chr.11:86441108-86674745 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ME3 | NM_001014811.1 | 9 | 1334 | NP_001014811.1 |
NM_001161586.1 | 10 | 1407 | NP_001155058.1 | |
NM_006680.2 | 10 | 1182 | NP_006671.2 | |
XM_005273717.1 | 10 | 1283 | XP_005273774.1 | |
XM_017017136.1 | 10 | 2165 | XP_016872625.1 | |
XM_017017137.1 | 6 | 707 | XP_016872626.1 | |
XM_017017138.1 | 11 | 1291 | XP_016872627.1 | |
AA403046.1 | 5 | 422 | ||
AA449933.1 | ||||
AK300974.1 | ||||
AK313695.1 | 10 | 1257 | ||
BC022472.2 | 9 | 1334 | AAH22472.1 | |
BQ882896.1 | 1 | 67 | ||
X79440.1 | 9 | 1112 | CAA55956.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |