Assay Details
Target Gene Details
Entrez Gene ID: | 23659 |
Gene Name: | phospholipase A2 group XV |
Gene Aliases: |
ACS, GXVPLA2, LLPL, LPLA2, LYPLA3 |
Location: |
Chr.16:68245344-68261062 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLA2G15 | NM_012320.3 | NP_036452.1 | ||
XM_005255886.3 | XP_005255943.1 | |||
XM_011522979.2 | XP_011521281.1 | |||
XM_011522980.2 | XP_011521282.2 | |||
AB017494.1 | BAA76877.1 | |||
AK001705.1 | ||||
AK074828.1 | BAC11233.1 | |||
AK127898.1 | 1 | 1004 | ||
AK222790.1 | BAD96510.1 | |||
AK296263.1 | ||||
AK298494.1 | ||||
AK300596.1 | ||||
AL110209.1 | CAB53675.1 | |||
AY358425.1 | AAQ88791.1 | |||
BC011640.2 | AAH11640.2 | |||
BC062605.1 | AAH62605.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |