Assay Details
Target Gene Details
Entrez Gene ID: | 4152 |
Gene Name: | methyl-CpG binding domain protein 1 |
Gene Aliases: |
CXXC3, PCM1, RFT |
Location: |
Chr.18:50266882-50281774 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv576889 | Chr.18:50227099 - 50287465 on Build GRCh38 | Loss | CFAP53 MBD1 CXXC1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |