Assay Details
Target Gene Details
Entrez Gene ID: | 111 |
Gene Name: | adenylate cyclase 5 |
Gene Aliases: |
AC5, FDFM |
Location: |
Chr.3:123282296-123448632 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 29 - Exon 29 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADCY5 | NM_001199642.1 | NP_001186571.1 | ||
NM_183357.2 | NP_899200.1 | |||
XM_005247077.3 | XP_005247134.1 | |||
XM_005247078.1 | XP_005247135.1 | |||
XM_006713483.1 | XP_006713546.1 | |||
XM_006713484.1 | XP_006713547.1 | |||
XM_011512359.2 | XP_011510661.1 | |||
XM_011512360.2 | XP_011510662.1 | |||
XM_011512361.1 | XP_011510663.1 | |||
XM_017005638.1 | XP_016861127.1 | |||
XM_017005639.1 | XP_016861128.1 | |||
XM_017005640.1 | XP_016861129.1 | |||
AF497517.1 | AAM94374.1 | |||
AK093840.1 | ||||
AK124691.1 | ||||
AK294565.1 | ||||
AK302711.1 | ||||
AK303070.1 | ||||
AK316488.1 | ||||
BX647739.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |