Assay Details
Target Gene Details
Entrez Gene ID: | 5832 |
Gene Name: | aldehyde dehydrogenase 18 family member A1 |
Gene Aliases: |
ADCL3, ARCL3A, GSAS, P5CS, PYCS, SPG9A, SPG9B |
Location: |
Chr.10:95605929-95656810 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ALDH18A1 | NM_001017423.1 | NP_001017423.1 | ||
NM_001323412.1 | NP_001310341.1 | |||
NM_001323413.1 | NP_001310342.1 | |||
NM_001323414.1 | NP_001310343.1 | |||
NM_001323415.1 | NP_001310344.1 | |||
NM_001323416.1 | NP_001310345.1 | |||
NM_001323417.1 | NP_001310346.1 | |||
NM_001323418.1 | NP_001310347.1 | |||
NM_001323419.1 | NP_001310348.1 | |||
NM_002860.3 | NP_002851.2 | |||
AK225455.1 | ||||
AK225465.1 | ||||
AK226032.1 | ||||
AK295487.1 | ||||
AK299557.1 | ||||
AK312271.1 | ||||
BC106054.1 | AAI06055.1 | |||
BC117240.1 | ||||
BC117242.1 | ||||
BC143930.1 | ||||
U68758.1 | AAD00169.1 | |||
U76542.1 | AAD17454.1 | |||
X94453.1 | CAA64224.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv526180 | Chr.10:95572438 - 95751280 on Build GRCh38 | Gain | ENTPD1 ALDH18A1 TCTN3 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |