Assay Details
Target Gene Details
Entrez Gene ID: | 8409 |
Gene Name: | ubiquitously expressed prefoldin like chaperone |
Gene Aliases: |
ART-27, STAP1 |
Location: |
Chr.X:47651792-47659180 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UXT | NM_004182.3 | NP_004173.1 | ||
NM_153477.2 | NP_705582.1 | |||
NR_045559.1 | ||||
NR_045560.1 | ||||
AF083241.1 | AAD39839.1 | |||
AF092737.1 | AAD28698.1 | |||
AK223320.1 | ||||
AK312072.1 | ||||
DA372561.1 | ||||
EL949569.1 |
Target Gene Details
Entrez Gene ID: | 100133957 |
Gene Name: | UXT antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.X:47658833-47660111 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UXT-AS1 | NR_028119.1 | |||
AK295794.1 | 1 | 320 | ||
DB182705.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |