Assay Details
Target Gene Details
Entrez Gene ID: | 6507 |
Gene Name: | solute carrier family 1 member 3 |
Gene Aliases: |
EA6, EAAT1, GLAST, GLAST1 |
Location: |
Chr.5:36606355-36688334 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC1A3 | NM_001166695.2 | 1 | 284 | NP_001160167.1 |
NM_001166696.2 | 1 | 284 | NP_001160168.1 | |
NM_001289939.1 | 1 | 284 | NP_001276868.1 | |
NM_001289940.1 | 1 | 284 | NP_001276869.1 | |
NM_004172.4 | 1 | 284 | NP_004163.3 | |
XM_005248342.2 | 1 | 49 | XP_005248399.1 | |
XM_011514084.2 | XP_011512386.1 | |||
AK057823.1 | ||||
AK293886.1 | ||||
AK295032.1 | ||||
AK295754.1 | ||||
AK312304.1 | 1 | 52 | ||
BC037310.2 | 1 | 52 | AAH37310.1 | |
BC054475.1 | AAH54475.1 | |||
D26443.1 | BAA05462.1 | |||
DC320625.1 | 1 | 284 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2762516 | Chr.5:36599283 - 36613099 on Build GRCh38 | Loss | SLC1A3 |
nsv830263 | Chr.5:36576402 - 36754991 on Build GRCh38 | Gain | SLC1A3 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |