Assay Details
Target Gene Details
Entrez Gene ID: | 10645 |
Gene Name: | calcium/calmodulin dependent protein kinase kinase 2 |
Gene Aliases: |
CAMKK, CAMKKB |
Location: |
Chr.12:121237692-121298308 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv560455 | Chr.12:121087788 - 121363295 on Build GRCh38 | Gain | ANAPC5 P2RX4 CAMKK2 P2RX7 |
nsv1054878 | Chr.12:121082077 - 121328424 on Build GRCh38 | Gain | ANAPC5 P2RX4 CAMKK2 P2RX7 |
nsv560454 | Chr.12:121019270 - 121307939 on Build GRCh38 | Loss | OASL P2RX4 CAMKK2 P2RX7 |
nsv455731 | Chr.12:121230451 - 121249371 on Build GRCh38 | Loss | P2RX4 CAMKK2 |
nsv521928 | Chr.12:120603925 - 121356945 on Build GRCh38 | Gain | HNF1A MLEC P2RX4 MIR4700 C12orf43 CAMKK2 CABP1 SPPL3 HNF1A-AS1 ANAPC5 OASL ACADS UNC119B P2RX7 XLOC_009911 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |