Assay Details
Target Gene Details
Entrez Gene ID: | 158038 |
Gene Name: | leucine rich repeat and Ig domain containing 2 |
Gene Aliases: |
LERN3, LRRN6C |
Location: |
Chr.9:27937617-29213981 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINGO2 | NM_001258282.1 | 7 | 2518 | NP_001245211.1 |
NM_152570.2 | 7 | 2598 | NP_689783.1 | |
XM_011517719.1 | 6 | 3466 | XP_011516021.1 | |
XM_011517724.2 | 5 | 6485 | XP_011516026.1 | |
XM_011517728.2 | 2 | 6264 | XP_011516030.1 | |
XM_017014303.1 | 11 | 6369 | XP_016869792.1 | |
XM_017014304.1 | 4 | 8889 | XP_016869793.1 | |
XM_017014305.1 | 5 | 8472 | XP_016869794.1 | |
XM_017014306.1 | 3 | 5633 | XP_016869795.1 | |
XM_017014307.1 | 2 | 3106 | XP_016869796.1 | |
AK056372.1 | 7 | 2598 | BAB71167.1 | |
AK172834.1 | 1 | 1598 | BAD18799.1 | |
AK290972.1 | 2 | 2129 | ||
AY358114.1 | 2 | 2152 | AAQ88481.1 | |
BC137514.1 | 7 | 2458 | ||
BC144676.1 | 7 | 2458 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv613950 | Chr.9:27746787 - 28342181 on Build GRCh38 | Gain | LINGO2 LOC646700 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |